5-Bromo-2-chloro-3-iodopyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-chloro-3-iodo-pyridine

A338016-1G A338016-25G A338016-5G A338016-100G A338016-10G A338016-250MG
103825-764EA 6 USD
103825-764 103836-734 76839-758 76839-760 76839-762 77015-510 77015-512 77015-514
5-Bromo-2-chloro-3-iodopyridine 98%
5-Bromo-2-chloro-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClIN
MW: 318.34 g/mol
Boiling Pt: 397.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08688585
CAS Number: 928653-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR