5-Bromo-2-chloro-3-fluorophenol 97%

Supplier: Ambeed
Warning

A156695-100MG A156695-250MG
103804-914EA 30 USD
103804-914 103843-116 77162-992 77162-994
5-Bromo-2-chloro-3-fluorophenol 97%
5-Bromo-2-chloro-3-fluorophenol

Precaución: Sealed in dry, at 2…8°C

Formula: C₆H₃BrClFO
MW: 225.443 g/mol
Boiling Pt: 241.5±35.0 °C (760 mm Hg)
Flash Pt: 99.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD27500640
CAS Number: 186590-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR