5-Bromo-2-chloro-1,3-difluorobenzene 97%

Supplier: Ambeed
Warning

A129202-100G A129202-10G A129202-1G A129202-25G A129202-5G
103801-242EA 9 USD
103801-242 77285-242 77285-244 77313-608 77313-610 77313-612
5-Bromo-2-chloro-1,3-difluorobenzene 97%
5-Bromo-2-chloro-1,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClF₂
Boiling Pt: 190.5 °C (760 mm Hg)
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00143420
CAS Number: 176673-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR