5-Bromo-2,4-dimethylpyridine ≥95%

Supplier: Ambeed
Warning

A898450-100MG A898450-250MG
103841-340EA 1246 USD
103841-340 77674-336 77884-846
5-Bromo-2,4-dimethylpyridine ≥95%
5-Bromo-2,4-dimethylpyridine

Precaución: Inert atmosphere, room temperature

Formula: C₇H₈BrN
Boiling Pt: 211.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09842454
CAS Number: 27063-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR