5-Bromo-2,4-dimethoxypyrimidine 98%

Supplier: Ambeed
Warning

A115673-100G A115673-10G A115673-1G A115673-25G A115673-5G
103824-462EA 7 USD
103824-462 76971-140 76971-142 76971-144 76971-146 77297-386
5-Bromo-2,4-dimethoxypyrimidine 98%
5-Bromo-2,4-dimethoxypyrimidine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O₂
Boiling Pt: 297 °C (760 mm Hg)
Melting Pt: 62…65 °C
Flash Pt: 133.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00038016
CAS Number: 56686-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR