1-Bromo-2,4-difluoro-5-nitrobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2,4-difluoronitrobenzene

A162835-10G A162835-1G A162835-25G
103805-662EA 12 USD
103805-662 103843-916 77150-970 77300-610 77300-612
1-Bromo-2,4-difluoro-5-nitrobenzene 97%
1-Bromo-2,4-difluoro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂NO₂
MW: 237.99 g/mol
Boiling Pt: 252.4 °C (760 mm Hg)
Flash Pt: 106.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00129166
CAS Number: 345-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR