5-Bromo-2,4-difluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A217675-100G A217675-25G A217675-10G
103818-634EA 7 USD
103818-634 103849-364 77013-174 77087-990 77683-627
5-Bromo-2,4-difluorobenzaldehyde 98%
5-Bromo-2,4-difluorobenzaldehyde

Precaución: For research use only.

Formula: C₇H₃BrF₂O
MW: 220.9989 g/mol
Boiling Pt: 229.4±35.0 °C (760 mm Hg)
Flash Pt: 92.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09842439
CAS Number: 473416-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR