5-Bromo-2,4-dichlorophenol 96%

Supplier: Ambeed
Warning

A328021-100MG A328021-250MG
103825-488EA 70 USD
103825-488 77032-842 77032-844
5-Bromo-2,4-dichlorophenol 96%
5-Bromo-2,4-dichlorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂O
MW: 241.89 g/mol
Boiling Pt: 277.5±35.0 °C (760 mm Hg)
Flash Pt: 121.6 °C
Storage Temperature: Ambient
MDL Number: MFCD16038723
CAS Number: 183803-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR