5-Bromo-2,3-dimethylpyridine 95%

Supplier: Ambeed
Warning

A387216-1G A387216-25G A387216-5G A387216-100G A387216-10G A387216-500G
103826-818EA 7 USD
103826-818 103837-860 76841-216 76841-218 76841-220 77033-514 77033-516 77033-518
5-Bromo-2,3-dimethylpyridine 95%
5-Bromo-2,3-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.0491 g/mol
Boiling Pt: 216.1±35.0 °C (760 mm Hg)
Flash Pt: 84.5 °C
Storage Temperature: Ambient
MDL Number: MFCD11656270
CAS Number: 27063-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR