5-Bromo-2,3-dimethylpyrazine 97%
Supplier: Ambeed
|
Warning
|
A126983-100MG
A126983-250MG
A126983-1G
103800-894EA
239
USD
103800-894
76822-756
76822-758
77313-142
5-Bromo-2,3-dimethylpyrazine 97%
5-Bromo-2,3-dimethylpyrazine
Precaución: for Research Use Only
Formula:
C₆H₇BrN₂ MW: 187.04 g/mol Boiling Pt: 218.0±35.0 °C (760 mm Hg) Flash Pt: 85.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD12404946 CAS Number: 111454-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...