5-Bromo-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-2,3-dihydro-7-azaindole
A141909-100MG
A141909-10G
A141909-250MG
A141909-1G
A141909-25G
A141909-5G
103803-076EA
13
USD
103803-076
77150-106
77150-108
77150-110
77316-274
77316-276
77316-278
5-Bromo-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 98%
5-Bromo-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only
Formula:
C₇H₇BrN₂ MW: 199.0479 g/mol Melting Pt: 186…191 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659750 CAS Number: 115170-40-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...