5-Bromo-1-indanone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-2,3-dihydro-1H-inden-1-one
A152552-100G
A152552-10G
A152552-1G
A152552-500G
A152552-5G
A152552-25G
103842-574EA
6
USD
103842-574
77259-292
77259-294
77259-296
77259-298
77299-958
77318-194
5-Bromo-1-indanone 98%
5-Bromo-1-indanone
Precaución: for Research Use Only
Formula:
C₉H₇BrO MW: 211.06 g/mol Boiling Pt: 303.7 °C (760 mm Hg) Melting Pt: 126…129 °C Flash Pt: 120.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082718 CAS Number: 34598-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...