5-Bromo-2,3-difluoropyridine 98%

Supplier: Ambeed
Warning

A136394-100G A136394-10G A136394-1G A136394-25G A136394-5G
103802-298EA 6 USD
103802-298 77109-568 77109-570 77315-182 77315-184 77315-186
5-Bromo-2,3-difluoropyridine 98%
5-Bromo-2,3-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrF₂N
MW: 193.98 g/mol
Boiling Pt: 164.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06659525
CAS Number: 89402-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR