5-Bromo-2,3-difluorobenzaldehyde 95%

Supplier: Ambeed
Warning

A244927-100MG A244927-250MG A244927-1G A244927-25G A244927-5G
103811-614EA 15 USD
103811-614 103851-448 76826-820 76826-822 77090-674 77090-676 77090-678
5-Bromo-2,3-difluorobenzaldehyde 95%
5-Bromo-2,3-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
MW: 220.999 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08669446
CAS Number: 633327-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR