5-Bromo-2,3-dichloropyridine 98%

Supplier: Ambeed
Warning

A237523-100G A237523-10G A237523-1G A237523-25G A237523-5G
103811-156EA 6 USD
103811-156 103850-934 76826-660 76826-662 77090-020 77090-022 77090-024
5-Bromo-2,3-dichloropyridine 98%
5-Bromo-2,3-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrCl₂N
MW: 226.89 g/mol
Boiling Pt: 233.6 °C (760 mm Hg)
Melting Pt: 35…37 °C
Flash Pt: 95.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04112492
CAS Number: 97966-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR