5-Bromo-2,3-diaminopyridine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromopyridine-2,3-diyldiamine, 2,3-Diamino-5-bromopyridine
A164323-10G
A164323-1G
A164323-25G
A164323-5G
A164323-100G
A164323-500G
103805-846EA
5
USD
103805-846
103844-106
76973-716
77320-264
77320-266
77320-268
77592-996
77671-318
5-Bromo-2,3-diaminopyridine 95%
5-Bromo-2,3-diaminopyridine
Precaución: for Research Use Only
Formula:
C₅H₆BrN₃ MW: 188.03 g/mol Boiling Pt: 321 Melting Pt: 155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00460094 CAS Number: 38875-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...