5-Bromo-2,2-difluoro-1,3-benzodioxole 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2,2-difluorobenzodioxole, 5-bromo-2,2-difluoro-2H-1,3-benzodioxole

A128809-100G A128809-10G A128809-500G A128809-5G A128809-1G A128809-25G
103801-184EA 5 USD
103801-184 77109-072 77109-074 77109-076 77109-078 77313-536 77313-538
5-Bromo-2,2-difluoro-1,3-benzodioxole 97%
5-Bromo-2,2-difluoro-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O₂
MW: 237 g/mol
Boiling Pt: 187.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00236212
CAS Number: 33070-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR