5-Bromo-2,2'-bithiophene 97%

Supplier: Ambeed
Warning

A231207-10G A231207-250MG A231207-1G
103810-762EA 25 USD
103810-762 103850-510 76771-158 76771-160 77089-390
5-Bromo-2,2'-bithiophene 97%
5-Bromo-2,2'-bithiophene

Precaución: for Research Use Only

Formula: C₈H₅BrS₂
MW: 245.16 g/mol
Boiling Pt: 286 °C (760 mm Hg)
Melting Pt: 29…32 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01461947
CAS Number: 3480-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR