5-Bromo[2,2']bipyridinyl 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2,2'-bipyridine

A101337-100MG A101337-10G A101337-250MG A101337-25G A101337-1G A101337-5G
103795-916EA 22 USD
103795-916 103821-602 76765-158 76765-160 76765-162 76765-164 77297-058 77297-060
5-Bromo[2,2']bipyridinyl 98%
5-Bromo[2,2']bipyridinyl

Precaución: for Research Use Only

Formula: C₁₀H₇BrN₂
MW: 235.09 g/mol
Flash Pt: 148.8 °C
Storage Temperature: Ambient
MDL Number: MFCD08234848
CAS Number: 15862-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR