5-Bromo-2-(trifluoromethyl)pyrimidine 96%

Supplier: Ambeed
Warning

A162154-250MG A162154-1G A162154-25G A162154-5G
103805-580EA 14 USD
103805-580 103843-830 76783-910 77319-866 77319-868 77319-870
5-Bromo-2-(trifluoromethyl)pyrimidine 96%
5-Bromo-2-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₂BrF₃N₂
MW: 226.98 g/mol
Boiling Pt: 139.8 °C (760 mm Hg)
Melting Pt: 41…42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09754046
CAS Number: 799557-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR