5-Bromo-2-(trifluoromethyl)phenol 95%

Supplier: Ambeed
Warning

A119603-100MG A119603-250MG
103799-798EA 174 USD
103799-798 76983-162 76983-164
5-Bromo-2-(trifluoromethyl)phenol 95%
5-Bromo-2-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O
Storage Temperature: Refrigerator
MDL Number: MFCD07787469
CAS Number: 1121585-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR