5-Bromo-2-(trifluoromethoxy)benzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-cyano-1-(trifluoromethoxy)benzene

A125266-100G A125266-10G A125266-250MG A125266-25G A125266-1G A125266-5G
103800-658EA 19 USD
103800-658 77026-734 77026-736 77026-738 77026-740 77312-808 77312-810
5-Bromo-2-(trifluoromethoxy)benzonitrile 97%
5-Bromo-2-(trifluoromethoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃BrF₃NO
MW: 266.015 g/mol
Boiling Pt: 244 °C (760 mm Hg)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD13185875
CAS Number: 1210906-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR