5-Bromo-2-(methylthio)pyrimidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A112032-100G A112032-10G A112032-1G A112032-5G
103797-938EA 5 USD
103797-938 103823-766 76970-888 76970-890 77284-202 77310-576
5-Bromo-2-(methylthio)pyrimidine-4-carboxylic acid 97%
5-Bromo-2-(methylthio)pyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂S
MW: 249.0851 g/mol
Boiling Pt: 414.5 °C (760 mm Hg)
Melting Pt: 158…162 °C
Flash Pt: 204.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00238866
CAS Number: 50593-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR