5-Bromo-2-(bromomethyl)benzonitrile 95%

Supplier: Ambeed
Danger

A103615-100MG A103615-5G
103796-578EA 272 USD
103796-578 103822-284 77148-326 77309-160
5-Bromo-2-(bromomethyl)benzonitrile 95%
5-Bromo-2-(bromomethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₅Br₂N
MW: 274.94 g/mol
Boiling Pt: 307.2±27.0 °C (760 mm Hg)
Flash Pt: 139.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09261033
CAS Number: 156001-53-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR