5-Bromo-1-methylisatin 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1-methylindoline-2,3-dione

A394784-1G A394784-100MG A394784-250MG A394784-5G
103826-950EA 23 USD
103826-950 103837-988 76841-410 77016-250 77016-252 77307-034
5-Bromo-1-methylisatin 95%
5-Bromo-1-methylisatin

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MW: 240.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00456313
CAS Number: 2058-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR