5-Bromo-1-methyl-1H-indazole 98%

Supplier: Ambeed
Warning

A115314-100G A115314-10G A115314-250MG A115314-25G A115314-1G A115314-5G
103798-542EA 6 USD
103798-542 103824-396 76781-352 76781-354 76781-356 76781-358 77311-206 77311-208
5-Bromo-1-methyl-1H-indazole 98%
5-Bromo-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Boiling Pt: 293.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09878568
CAS Number: 465529-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR