5-Bromo-1-methylimidazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-1-methyl-1H-imidazole
A182178-100G
A182178-10G
A182178-500G
A182178-25G
A182178-5G
A182178-1G
103807-688EA
6
USD
103807-688
103846-060
77012-032
77012-034
77012-036
77083-596
77083-598
77301-772
5-Bromo-1-methylimidazole 97%
5-Bromo-1-methylimidazole
Precaución: for Research Use Only
Formula:
C₄H₅BrN₂ MW: 161 g/mol Boiling Pt: 110 °C (760 mm Hg) Melting Pt: 40…44 °C Flash Pt: 82.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632218 CAS Number: 1003-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...