5-Bromo-1-methyl-1H-benzo[d][1,2,3]triazole 97%

Supplier: Ambeed
Warning

A371670-100MG A371670-250MG A371670-1G A371670-5G
103826-488EA 235 USD
103826-488 103837-506 77016-004 77016-006 77306-836 77306-838
5-Bromo-1-methyl-1H-benzo[d][1,2,3]triazole 97%
5-Bromo-1-methyl-1H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Boiling Pt: 332.6±15.0 °C (760 mm Hg)
Flash Pt: 154.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19288764
CAS Number: 944718-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR