5-Bromo-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A166254-100MG
A166254-10G
A166254-250MG
A166254-25G
A166254-1G
A166254-5G
103806-038EA
36
USD
103806-038
103844-298
76813-012
76813-014
76813-016
76813-018
77320-542
77320-544
5-Bromo-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid 98%
5-Bromo-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₅BrN₂O₂ MW: 241.0415 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12024580 CAS Number: 800401-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...