5-Bromo-6-azaindole 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1H-pyrrolo[2,3-c]pyridine

A113356-100MG A113356-250MG A113356-1G A113356-5G
103798-218EA 17 USD
103798-218 103824-060 76810-044 76810-046 77310-874 77310-876
5-Bromo-6-azaindole 98%
5-Bromo-6-azaindole

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
Storage Temperature: Ambient
MDL Number: MFCD12024306
CAS Number: 1215387-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR