5-Bromo-1H-pyrrolo[2,3-c]pyridin-2(3H)-one 95%

Supplier: Ambeed
Warning

A102669-100MG A102669-250MG A102669-5G A102669-1G
103796-404EA 310 USD
103796-404 76809-354 76809-356 76809-358 77282-952
5-Bromo-1H-pyrrolo[2,3-c]pyridin-2(3H)-one 95%
5-Bromo-1H-pyrrolo[2,3-c]pyridin-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
MW: 213.031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848386
CAS Number: 1217002-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR