5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-7-azaindole-3-carboxylic acid
A379370-100MG
A379370-250MG
A379370-1G
A379370-5G
103826-646EA
49
USD
103826-646
103837-682
76773-564
76773-566
76840-990
76840-992
5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid 97%
5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₅BrN₂O₂ MW: 241.0400 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09965908 CAS Number: 849068-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...