5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 97%

Supplier: Ambeed
Warning

A210999-250MG A210999-1G
103818-064EA 15 USD
103818-064 103848-756 76975-554 77087-154
5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 97%
5-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD09755969
CAS Number: 757978-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR