5-Bromo-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-7-azaindole

A106975-100G A106975-10G A106975-250MG A106975-25G A106975-500G A106975-1G A106975-5G
103797-102EA 9 USD
103797-102 103822-860 76994-960 76994-962 76994-964 76994-966 76994-968 77283-522 77309-728
5-Bromo-1H-pyrrolo[2,3-b]pyridine 98%
5-Bromo-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
Melting Pt: 178.0…178.8 °C
Storage Temperature: Ambient
MDL Number: MFCD06659677
CAS Number: 183208-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR