5-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1H-pyrrolo[2,3-b]pyridin-2-one

A205059-250MG A205059-25G A205059-1G A205059-5G
103817-586EA 77 USD
103817-586 103848-250 76770-300 76770-302 77086-450 77302-932
5-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 97%
5-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
MW: 213.0300 g/mol
Boiling Pt: 387.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07367221
CAS Number: 183208-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR