5-Bromo-1H-pyrazolo[3,4-c]pyridine 98%
Supplier: Ambeed
|
Warning
|
A237252-10G
A237252-250MG
A237252-1G
A237252-5G
A237252-25G
103811-140EA
11
USD
103811-140
103850-918
76786-426
76786-428
77089-990
77089-992
77304-078
5-Bromo-1H-pyrazolo[3,4-c]pyridine 98%
5-Bromo-1H-pyrazolo[3,4-c]pyridine
Precaución: for Research Use Only
Formula:
C₆H₄BrN₃ MW: 198.0201 g/mol Flash Pt: 186 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11518975 CAS Number: 929617-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...