5-Bromo-1H-pyrazolo[3,4-b]pyridine 97%

Supplier: Ambeed
Warning

A217103-100G A217103-10G A217103-250MG A217103-500G A217103-25G A217103-5G A217103-1G
103818-600EA 8 USD
103818-600 103849-328 77013-162 77013-164 77013-166 77013-168 77087-942 77087-944 77303-446
5-Bromo-1H-pyrazolo[3,4-b]pyridine 97%
5-Bromo-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.02 g/mol
Flash Pt: 152.8 °C
Storage Temperature: Ambient
MDL Number: MFCD05663982
CAS Number: 875781-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR