5-Bromo-1H-pyrazolo[3,4-b]pyrazine 94%
Supplier: Ambeed
|
Warning
|
A168203-100MG
A168203-250MG
A168203-1G
103806-262EA
407
USD
103806-262
76872-280
76872-282
77300-922
5-Bromo-1H-pyrazolo[3,4-b]pyrazine 94%
5-Bromo-1H-pyrazolo[3,4-b]pyrazine
Precaución: for Research Use Only
Formula:
C₅H₃BrN₄ MW: 199.01 g/mol Boiling Pt: 333.8 °C (760 mm Hg) Flash Pt: 155.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD13190158 CAS Number: 1196152-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...