5-Bromo-1H-pyrazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A130867-100MG A130867-250MG A130867-1G
103801-486EA 25 USD
103801-486 76811-178 76811-180 77313-956
5-Bromo-1H-pyrazole-3-carboxylic acid 97%
5-Bromo-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₃BrN₂O₂
MW: 190.983 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21605341
CAS Number: 1328893-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR