5-Bromo-1H-pyrazole 95%

Supplier: Ambeed
Warning

A119476-10G A119476-250MG A119476-25G A119476-1G A119476-5G
103799-772EA 18 USD
103799-772 77009-366 77009-368 77009-370 77297-686 77297-688
5-Bromo-1H-pyrazole 95%
5-Bromo-1H-pyrazole

Precaución: for Research Use Only

Formula: C₃H₃BrN₂
MW: 146.97 g/mol
Boiling Pt: 70…72 °C (0.1 mm Hg)
Melting Pt: 70…72 °C
Flash Pt: 113.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13189492
CAS Number: 1174132-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR