5-Bromo-1H-indole-7-carboxylic acid 97%

Supplier: Ambeed
Warning

A268689-1G A268689-250MG
103812-930EA 72 USD
103812-930 103852-832 77092-592 77612-574
5-Bromo-1H-indole-7-carboxylic acid 97%
5-Bromo-1H-indole-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MW: 240.053 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845481
CAS Number: 860624-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR