5-Bromo-1H-indole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A155076-1G A155076-5G
103804-716EA 8 USD
103804-716 103842-912 77010-952 77010-954 77318-640 77318-642
5-Bromo-1H-indole-3-carboxylic acid 97%

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
Boiling Pt: 215.8 °C (760 mm Hg)
Flash Pt: 238.6 °C
Storage Temperature: Ambient
MDL Number: MFCD05664007
CAS Number: 10406-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR