5-Bromoindole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1H-indole-2-carboxylic acid

A182595-5G A182595-100G A182595-10G A182595-250MG
103807-730EA 20 USD
103807-730 103846-104 77083-650 77274-132 77615-542 77615-544
5-Bromoindole-2-carboxylic acid 98%
5-Bromoindole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MW: 240.06 g/mol
Boiling Pt: 470.9 °C (760 mm Hg)
Melting Pt: 287…288 °C
Flash Pt: 238.611 °C
Storage Temperature: Freezer
MDL Number: MFCD00022705
CAS Number: 7254-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR