5-Bromo-1H-indole-2-carbonitrile 95%

Supplier: Ambeed
Warning

A299313-100MG A299313-250MG A299313-5G
103814-220EA 262 USD
103814-220 103835-366 76827-836 76827-838 77305-762
5-Bromo-1H-indole-2-carbonitrile 95%
5-Bromo-1H-indole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₅BrN₂
MW: 221.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20274509
CAS Number: 902772-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR