5-Bromo-1H-indol-6-amine 97%
Supplier: Ambeed
|
Warning
|
A396544-100MG
A396544-250MG
103826-986EA
193
USD
103826-986
76773-694
76773-696
5-Bromo-1H-indol-6-amine 97%
5-Bromo-1H-indol-6-amine
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂ MW: 211.06 g/mol Boiling Pt: 387 °C (760 mm Hg) Flash Pt: 187.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09880057 CAS Number: 873055-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...