5-Bromo-1H-inden-2(3H)-one 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromoindan-2-one

A210146-100MG A210146-250MG
103817-980EA 83 USD
103817-980 103848-672 77261-008 77261-010
5-Bromo-1H-inden-2(3H)-one 97%
5-Bromo-1H-inden-2(3H)-one

Precaución: for Research Use Only

Formula: C₉H₇BrO
MW: 211.0600 g/mol
Boiling Pt: 353.8 °C (760 mm Hg)
Melting Pt: 123…127 °C
Flash Pt: 117.2 °C
Storage Temperature: Ambient
MDL Number: MFCD07778391
CAS Number: 174349-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR