5-Bromo-1H-indazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A247262-10G A247262-250MG A247262-1G A247262-25G A247262-5G
103811-760EA 6 USD
103811-760 103851-594 77000-598 77000-600 77090-920 77090-922 77090-924
5-Bromo-1H-indazole-3-carboxylic acid 97%
5-Bromo-1H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
MW: 241.0400 g/mol
Boiling Pt: 493.4 °C (760 mm Hg)
Melting Pt: 289…292 °C
Flash Pt: 252.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05663979
CAS Number: 1077-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR