5-Bromo-1H-indazole-3-carbonitrile 97%

Supplier: Ambeed
Warning

A213717-100MG A213717-10G A213717-250MG A213717-1G
103818-320EA 95 USD
103818-320 103849-028 77013-024 77013-026 77013-028 77303-306
5-Bromo-1H-indazole-3-carbonitrile 97%
5-Bromo-1H-indazole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrN₃
Boiling Pt: 425.3 °C (760 mm Hg)
Flash Pt: 211.0 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07371567
CAS Number: 201227-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR