5-Bromo-1H-indazole-3-carbaldehyde 97%

Supplier: Ambeed
Warning

A383002-1G A383002-5G A383002-10G A383002-250MG
103826-728EA 14 USD
103826-728 103837-764 76841-084 76841-086 77016-086 77016-088
5-Bromo-1H-indazole-3-carbaldehyde 97%
5-Bromo-1H-indazole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
MW: 225.042 g/mol
Boiling Pt: 414 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06738287
CAS Number: 201227-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR