5-Bromo-1H-indazole 98%

Supplier: Ambeed
Warning

A212006-100G A212006-10G A212006-500G A212006-1G A212006-25G A212006-5G
103818-154EA 5 USD
103818-154 103848-850 76814-570 76814-572 76814-574 77087-286 77087-288 77087-290
5-Bromo-1H-indazole 98%
5-Bromo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.03 g/mol
Boiling Pt: 333.8 °C (760 mm Hg)
Melting Pt: 131…135 °C
Flash Pt: 155.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00839493
CAS Number: 53857-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR